Order Entry
United States
ContactUsLinkComponent
 
Mouse Recombinant GITRL (soluble) (from HEK293 cells)
Catalog # 102514-742
Supplier:  Adipogen
Small Business Enterprise
CAS Number:  
Mouse Recombinant GITRL (soluble) (from HEK293 cells)
Catalog # 102514-742
Supplier:  Adipogen
Supplier Number:  AG-40A-0008-C050
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Size:
    50 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.1EU/µg
  • Biological Activity:
    Induces GITR-mediated NF-kappaB activation in HEK 293 cells.
  • Gene ID:
    Q7TS55
  • Protein Synonyms:
    Activation-inducible TNF-related Ligand|AITRL|TNFSF18|Glucocorticoid-induced TNF Receptor Ligand
  • Protein/Peptide Name:
    GITRL (soluble)
  • Purity:
    90%
  • Molecular Weight:
    ~22kDa (SDS-PAGE)
  • Sequence:
    45-173
  • Endotoxin Level:
    Low
  • Concentration:
    0.5mg/ml
  • Formulation:
    Liquid. 0.2µm-filtered solution in 10mM glycine, pH 10.0.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102514-742
  • Supplier no.:
    AG40A0008C050

Specifications

About this item

GITRL (Glucocorticoid-induced TNF receptor ligand) is expressed on dendritic cells (DC), monocytes, macrophages, B cells, activated T cells, endothelial cells, osteoclasts and various healthy non-lymphoid tissues (e.g. testis). GITRL is constitutively expressed and released as soluble form by solid tumors and various hematopoietic malignancies. GITRL causes differentiation of osteoclasts, activation of macrophages, but also alteration of carcinoma and leukemia cells and influences apoptosis. Binding to GITR is important in regulating T cell proliferation and TCR-mediated apoptosis. GITRL is implicated in development of autoimmune diseases and in the immune response against infectious pathogens and tumors.