Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:A Proliferation-inducing Ligand
- Antigen Symbol:APRIL
- Clone:Apry-1-1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:HEK293
- ImmunoPrecipitation:Yes
- Isotype:IgG2b lambda
- Reactivity:Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:Q9D777
- Antigen Synonyms:TNFSF13|A Proliferation Inducing Ligand|TALL-2|CD256
- Storage Buffer:In PBS containing 10% glycerol.
- Storage Temperature:Short term: +4 °C, Long Term: –20 °C
- Concentration:1mg/ml
- Shipping Temperature:-20° C
- Immunogen:Mouse APRIL (aa 98-232).
- Tested Applications:Functional Blocking
- Cat. No.:102511-416
- Supplier no.:27B0001PFC100
Specifications
About this item
APRIL is a cytokine that belongs to the TNF superfamily and binds to TACI and BCMA. It is implicated in the regulation of tumor cell growth and is involved in monocyte/macrophage-mediated immunological processes. Anti-APRIL (mouse) Monoclonal Antibody (recombinant) (Blocking) (APRY-1-1) is an antibody developed by antibody phage display technology using a human naive antibody gene library. These libraries consist of scFv (single chain fragment variable) composed of VH (variable domain of the human immunoglobulin heavy chain) and VL (variable domain of the human immunoglobulin light chain) connected by a polypeptide linker. The antibody fragments are displayed on the surface of filamentous bacteriophage (M13). This scFv was selected by affinity selection on antigen in a process termed panning. Multiple rounds of panning are performed to enrich for antigen-specific scFv-phage. Monoclonal antibodies are subsequently identified by screening after each round of selection. The selected monoclonal scFv is cloned into an appropriate vector containing a Fc portion of interest and then produced in mammalian cells to generate an IgG like scFv-Fc fusion protein.
- Stable for at least 1 year after receipt when stored at -20°C.
Purified from HEK 293 cell culture supernatant.
Purity: >95% (SDS-PAGE)
Type:
Antigen: APRIL
Clonality:
Clone: Apry-1-1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b lambda
Reactivity: Mouse