Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:Cornulin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9UBG3
- Antigen Synonyms:Chromosome 1 Open Reading Frame 10 Gene Product|C1orf10 Gene Product|Tumor Related Protein
- Storage Buffer:In PBS containing 1% BSA and 0.01% sodium azide.
- Storage Temperature:Short term: +4 °C, Long Term: –20 °C
- Concentration:0.5mg/ml
- Shipping Temperature:-20° C
- Immunogen:Synthetic peptide corresponding to aa 259-274 (E259ATNDQNRGTETHGQG274) of human cornulin.
- Cat. No.:102514-674
- Supplier no.:AG25B0016C050
Specifications
About this item
Cornulin is a survival factor that participates in the clonogenicity of squamous esophageal epithelium cell lines. It attenuates deoxycholic acid (DCA)-induced apoptotic cell death and the release of calcium. In overexpressed oral squamous carcinoma cell lines, cornulin negatively regulates cell proliferation by the induction of G1 arrest.
Stable for at least 1 year after receipt when stored at -20°C.
Type:
Antigen: Cornulin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human