Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:ST2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q01638
- Antigen Synonyms:IL-1 Receptor-like 1|IL-33 Receptor|Interleukin-33 Receptor|Interleukin-1 Receptor-like 1|IL1RL1
- Storage Buffer:0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives.
- Storage Temperature:Short term: +4 °C, Long Term: –20 °C
- Concentration:1mg/ml
- Shipping Temperature:-20° C
- Immunogen:Recombinant human soluble ST2.
- Cat. No.:102514-522
- Supplier no.:AG25A0058C100
Specifications
About this item
The ST2 gene was originally identified as a gene induced by serum or oncogene expression in fibroblasts. The gene produces a shorter soluble secreted form (ST2) and a longer, transmembrane form (ST2L) by alternative splicing. Soluble ST2 has been shown to downregulate the expression of TLR1 and TLR4. ST2L negatively regulates TLR4 signaling and induces endotoxin tolerance, and enhances Th2 responses. IL-33 is the specific ligand for ST2L.
Type:
Antigen: ST2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human