Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Angiotensin Converting Enzyme 2
- Antigen Symbol:ACE2
- Clonality:Monoclonal
- Clone:AC18F
- Conjugation:Biotin
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9BYF1
- Antigen Synonyms:ACEH|Angiotensin-converting Enzyme 2|SARS Receptor
- Storage Buffer:0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives.
- Storage Temperature:Short term: +4 °C, Long Term: –20 °C
- Concentration:1mg/ml
- Shipping Temperature:-20° C
- Immunogen:Recombinant human ACE2.
- Tested Applications:FACS
- Cat. No.:102514-036
- Supplier no.:AG20A0032BC050
Specifications
About this item
ACE2 has direct effects on cardiac functions. It is expressed predominantly in vascular endothelial cells of the heart and kidney. ACE2 converts angiotensin I to angiotensin 1-9. ACE2 was also identified as a functional receptor for SARS coronavirus.
Type:
Antigen: ACE2
Clonality: Monoclonal
Clone: AC18F
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human