About this item
Tim-3 is a Th1-specific cell surface protein. It is expressed selectively by differentiated CD4+Th1 and CD8+Tc1 cells, but is absent on CD4+Th2 and CD8+ Tc2 cells. The Tim gene family has an important role in the regulation of autoimmunity and allergies.
Type:
Antigen: Tim-3
Clonality: Monoclonal
Clone: TI 339H
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- 102514-022
- 102514-020
- Antigen Symbol:
- TIM3
- TIM3
- Antigen Synonyms:
- TIMD3|Hepatitis A Virus Cellular Receptor 2|HAVcr-2|T Cell Immunoglobulin and Mucin Domain-containing Protein 3|TIM3
- TIMD3|Hepatitis A Virus Cellular Receptor 2|HAVcr-2|T Cell Immunoglobulin and Mucin Domain-containing Protein 3|TIM3
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- TI 339H
- TI 339H
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- Q8VIM0
- Q8VIM0
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant mouse Tim-3.
- Recombinant mouse Tim-3.
- Form:
- Liquid
- Liquid
- Concentration:
- 1mg/ml
- 1mg/ml
- Storage Buffer:
- 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives.
- 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives.
- Storage Temperature:
- Short term: +4 °C, Long Term: –20 °C
- Short term: +4 °C, Long Term: –20 °C
- Shipping Temperature:
- -20° C
- -20° C