Yeast Peptone Dextrose (YPD) Agar, Hardy Diagnostics
Catalog # 89407-316
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Plate
- Size:15 x 100 mm
- Environmentally Preferable:
- Storage Temperature:2-8°C
- Minimum Order Quantity:10
- Cat. No.:89407-316
- Volume:26 ml
- Supplier no.:G422
Specifications
About this item
CulGenex™ Yeast Peptone Dextrose (YPD) Media are recommended for the rapid growth of yeasts, particularly Saccharomyces cerevisiae, used for molecular research.