Saline, 0.85% with 0.05% Tween® 80, Hardy Diagnostics
Catalog # 89407-918
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Glass bottle, septum-sealed
- Size:100 mL
- Storage Temperature:–2-30°C
- Cat. No.:89407-918
- Supplier no.:U330
Specifications
About this item
For preparing dilutions and suspensions of microorganisms.