Order Entry
United States
ContactUsLinkComponent
PDX-LIB Media
PDX-LIB Media
Catalog # 89404-952
Supplier:  Avantor
Product available on GSA Advantage®
CAS Number:  
undefined
PDX-LIB Media
Catalog # 89404-952
Supplier:  Avantor
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Media Format:
    Liquid
  • Pack type:
    Tube
  • Description:
    PDX-LIB media
  • Size:
    25/case
  • Environmentally Preferable:
  • Storage Temperature:
    2...8 °C
  • Cat. No.:
    89404-952
  • Shelf life:
    1800
  • Application:
    Food safety
  • Volume:
    20 ml
  • Supplier no.:
    2500325

Specifications

About this item

Media for the detection of Listeria species from the environment, AOAC RI approved, 24 to 30 h result, closed system, no special equipment needed, use with EnviroMAX sampling sponge on a stick 89405-022.

This test is both reliable and inexpensive with presumptive results available for the most common Listeria spp, within 30 h, no sub-culturing or specimen transfers required. Simply collect your sample, inoculate the PDX-LIB media, and incubate. Detect as low as 10 to 50 heat injured Listeria monocytogenes per ml within 24 to 30 h of incubation. Comparable results to USDA methods. The test is AOAC-RI approved. Sampling devices are not included.