MUG Supplement, Hardy Diagnostics
Catalog # 89405-038
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Vial
- Size:10 vials
- Storage Temperature:2-8°C
- Cat. No.:89405-038
- Volume:50 mg
- Supplier no.:BR071E
Specifications
About this item
A fluorescent agent for the detection of Escherichia coli (contains 4-methylumbelifery-B-D-glucuronide 50 mg).