Specifications
- Pack type:Plate
- Size:99 mL
- Storage Temperature:2-8°C
- Minimum Order Quantity:10
- Cat. No.:89407-352
- Supplier no.:G86
Specifications
About this item
For the selective isolation and detection of Salmonella species Lysine Iron Agar (LIA) is recommended for use in differentiating certain members of the Enterobacteriaceae, especially Salmonella species, by demonstrating hydrogen sulfide production and the decarboxylation or deamination of lysine
Modified Lysine Iron Agar is recommended for the selective isolation and differentiation of Salmonella species in food, It contains novobiocin to inhibit the growth of gram-positive cocci.