Cetrimide Select Agar, Hardy Diagnostics
Catalog # 89407-198
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Plate
- Size:15×100 mm
- Storage Temperature:2-8°C
- Cat. No.:89407-198
- Volume:26 ml
- Supplier no.:G18
Specifications
About this item
For the selective isolation of Pseudomonas aeruginosa.