Order Entry
United States
ContactUsLinkComponent
Baird Parker Agar, Hardy Diagnostics
Baird Parker Agar, Hardy Diagnostics
Catalog # 89407-366
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Baird Parker Agar, Hardy Diagnostics
Catalog # 89407-366
Supplier:  Hardy Diagnostics
Supplier Number:  G96
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Pack type:
    Plate
  • Size:
    15 x 100 mm
  • Storage Temperature:
    2-8°C
  • Cat. No.:
    89407-366
  • Volume:
    18 ml
  • Supplier no.:
    G96

Specifications

About this item

For the selective isolation of coagulase positive Staphylococci.