ContactUsLinkComponent
Powerstat® Mounting Bracket, Ace Glass
Powerstat® Mounting Bracket, Ace Glass
Catalog # 89075-650
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Powerstat® Mounting Bracket, Ace Glass
Catalog # 89075-650
Supplier:  Ace Glass
Supplier Number:  11150-17
Product available on GSA Advantage®
Small Business Enterprise
Specifications
  • Description:
    Powerstat® Mounting Bracket for mounting of 12080 or 12081 Powerstats on standard laboratory racks or stands having up to 16 mm O.D. rods
  • Cat. No.:
    89075-650
  • Supplier no.:
    11150-17
Specifications
About this item
Mounting bracket permits easy and convenient mounting of Powerstat® on standard laboratory racks or stands having up to 16 mm O.D
Made of strong, rust- and corrosion-resistant aluminum alloys. Bracket is attached to the bottom of the Powerstat® by two screws and to the rack or stand by a twist of two thumbscrews.