ContactUsLinkComponent
Powerstat®, 0 - 280 Volts, Ace Glass Incorporated
  89076-324
 :  Ace Glass
Small Business Enterprise
undefined
Powerstat®, 0 - 280 Volts, Ace Glass Incorporated
  89076-324
 :  Ace Glass
 :  12082-10
Small Business Enterprise
  • Description:
    Powerstat® transformer
  • Environmentally Preferable:
  • Cat. No.:
    89076-324
  • Supplier no.:
    12082-10
Variable transformer for mantles, heating tops, heating cords, and heating tapes
Input is 230 volts, 50/60 cycles, single phase. Input is 1 PH, output is 0 – 280 volts, 10 amperes, 2.8 Kva. Fitted with standard NEMA three-blade plug and receptacle.