ContactUsLinkComponent
PIG® HazMat Spill Kit in Stowaway Bag, New Pig
PIG® HazMat Spill Kit in Stowaway Bag, New Pig
Catalog # 56620-426
Supplier:  New Pig
undefined
PIG® HazMat Spill Kit in Stowaway Bag, New Pig
Catalog # 56620-426
Supplier:  New Pig
Supplier Number:  KIT306
Specifications
  • Capacity:
    Up to 22.7 L (6 gal.)
  • Environmentally Preferable:
  • Cat. No.:
    56620-426
  • Supplier no.:
    KIT306
Specifications
About this item
Durable denim bag hides dirt and can fit almost anywhere
Temporary disposal bags help make cleanup easier. Spill kit in compact bag with drawstring closure is ideal for storage behind a vehicle seat.
PIG® HazMat socks stop spreading spills; PIG® HazMat pads absorb; PIG® repair putty plugs or patches cracks or leaks. PIG® HazMat absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide. Instant bucket helps you handle spills or ruptures on the road.