bisBenzimide H 33258 trihydrochloride (Hoechst 33258) ≥95% (by HPLC), Ultrapure dye
Catalog # 89166-028
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:100 mg
- Cat. No.:89166-028
- Supplier no.:52402
Specifications
About this item
:
:533.88 g/mol
Storage Temperature:Freezer
CAS Number:23491-45-4
Formula:C₂₅H₂₄N₆O·3HCl
MDL Number:MFCD00012679