Order Entry
United States
ContactUsLinkComponent
MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide) ≥98% (by HPLC), Ultrapure
MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide) ≥98% (by HPLC), Ultrapure
  89165-990
 :  
undefined
MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide) ≥98% (by HPLC), Ultrapure
  89165-990
 :  ENZ-52156
 :  

 

  • Size:
    100 mg
  • Cat. No.:
    89165-990
  • Supplier no.:
    52156

 

 

 :
 :326.19 g/mol
 :Freezer
 :162558-52-3
 :C₁₄H₁₆NO₃Br
 :MFCD00467854