Z-Gly-Gly-Leu-Amc
Catalog # 89165-650
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:5 mg
- Protein/Peptide Name:Z-Gly-Gly-Leu-AMC
- Purity:98%
- Cat. No.:89165-650
- Supplier no.:ZW8505-0005
Specifications
About this item
A fluorogenic substrate for the chymotrypsin-like activity of the proteasome.