Order Entry
United States
ContactUsLinkComponent
Human Recombinant Ubiquitin activating enzyme E1 (from E. coli), HIS Tag
Human Recombinant Ubiquitin activating enzyme E1 (from E. coli), HIS Tag
Catalog # 89165-374
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Human Recombinant Ubiquitin activating enzyme E1 (from E. coli), HIS Tag
Catalog # 89165-374
Supplier:  Enzo Life Sciences
Supplier Number:  UW9410-0050
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    HIS Tag
  • Protein/Peptide Name:
    Ubiquitin activating enzyme E1
  • Formulation:
    In 50mM TRIS-hydrochloric acid, containing 1.0mM dithiothreitol
  • Cat. No.:
    89165-374
  • Supplier no.:
    UW9410-0050

Specifications

About this item

Essential ubiquitinylation pathway enzyme.

Produced in E. coli. Full length human ubiquitin-activating enzyme E1.

Ubiquitin activating enzyme E1 is responsible for the first step in ubiquitin-protein isopeptide bond formation and is a critical component for the initiation of in vitro conjugation reactions. E1 activates ubiquitin by first adenylating with ATP the C-terminal glycine carboxyl group of ubiquitin and, thereafter, linking this residue to the sulphydryl side chain moiety of a cysteine residue in E1 by forming a high energy thiol ester bond and liberating free AMP. There are two active sites within the E1 molecule allowing it to accommodate two ubiquitin molecules at one time, with a new ubiquitin forming an adenylate intermediate as the previous one is transferred to the thiol site. The activated ubiquitin is then transferred to the lysine of target proteins via the E2/E3 conjugation cascade.