To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Melanocortin Receptor 2
- Antigen Symbol:MC2R
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:200 µL
- Cat. No.:89163-596
- Supplier no.:SA639-0200
Specifications
About this item
The melanocortin receptors are members of the G protein-coupled receptor superfamily. Their ligands, the melanocortins, are peptides consisting of α-, β- and γ-melanocyte stimulating hormone and adrenocorticotropic hormone. MC2R is only activated by ACTH. It is expressed gene is expressed in skin, scalp tissue, compound nevus tissue, basal cell carcinoma tissue, and cultured melanocytes.
- For Immunohistochemistry, Western Blot
Recognizes rat and human MC2R.
Type: Primary
Antigen: MC2R
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human