Anti-Tubulin Alpha Rabbit Polyclonal Antibody
Catalog # 89163-112
Supplier: Enzo Life Sciences
Specifications
- Antibody Type:Primary
- Antigen Name:Tubulin alpha
- Antigen Symbol:TUBA
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Primates
- Western Blot:Yes
- Size:100 µL
- Form:Liquid. In PBS.
- Antigen Synonyms:tubulin alpha-3 chain|tubulin B-alpha-1|alpha|tubulin|hum-a-tub1|alpha-tubulin 3|alpha 3|brain-specific|tubulin alpha-1A chain|hum-a-tub2
- Cat. No.:89163-112
- Supplier no.:SA452-0100
Specifications
About this item
Host: Rabbit
- For Western Blot
Recognizes primate, rodent, chicken, Xenopus and zebrafish alpha-tubulin. Detects a band of ~50kDa by Western blot.
Type: Primary
Antigen: TUBULIN ALPHA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Primate