To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Proteasome 19S Rpn8/S12 Subunit
- Antigen Symbol:Proteasome 19S Rpn8/S12
- Clonality:Polyclonal
- Host:Sheep
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:25 µL
- Form:Liquid. In PBS containing 10mM sodium azide.
- Cat. No.:89162-040
- Supplier no.:PW8180-0025
Specifications
About this item
Host: Sheep
- For Immunohistochemistry, Western Blot
Recognizes human proteasome 26S (19S regulator complex) (Rpn8/S12 subunit).
Type: Primary
Antigen: PROTEASOME 19S RPN8/S12 SUBUNIT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype:
Reactivity: Human