Order Entry
United States
ContactUsLinkComponent
Tetrodotoxin ≥98% (by TLC)
Tetrodotoxin ≥98% (by TLC)
Catalog # 89160-628
Supplier:  Avantor
CAS Number:  
Tetrodotoxin ≥98% (by TLC)
Catalog # 89160-628
Supplier:  Avantor
CAS Number:  

Specifications

  • Size:
    1 mg
  • Cat. No.:
    89160-628
  • Supplier no.:
    NA120-0001

Specifications

About this item

 :
 :319.27 g/mol
CAS Number:4368-28-9
Formula:C₁₁H₁₇N₃O₈
MDL Number:MFCD00213719