Ginkgolide B ≥90%
Catalog # 89160-478
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:10 mg
- Cat. No.:89160-478
- Supplier no.:L135-0010
Specifications
About this item
:
:424.40 g/mol
CAS Number:15291-77-7
Formula:C₂₀H₂₄O₁₀
MDL Number:MFCD05662347