Order Entry
United States
ContactUsLinkComponent
 
TCPOBOP ≥98% (by TLC)
  89158-844
 :  
TCPOBOP ≥98% (by TLC)
  89158-844
 :  BML-GR245-0025
 :  

 

  • Size:
    25 mg
  • Cat. No.:
    89158-844
  • Supplier no.:
    GR245-0025

 

 

  • Source:
    Synthetic
  • Purity:
    ≥98% (TLC)

 

 

 :
 :402.1 g/mol
 :Ambient
 :76150-91-9
 :C₁₆H₈Cl₄N₂O₂
 :MFCD00057368