Order Entry
United States
ContactUsLinkComponent
Sp-8-Br-cGMPS ≥98% (by HPLC)
Sp-8-Br-cGMPS ≥98% (by HPLC)
Catalog # 89157-046
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Sp-8-Br-cGMPS ≥98% (by HPLC)
Catalog # 89157-046
Supplier:  Enzo Life Sciences
Supplier Number:  BML-CN217-0500
CAS Number:  

Specifications

  • Size:
    500 µg
  • Cat. No.:
    89157-046
  • Supplier no.:
    CN217-0500

Specifications

Specification Test Results

  • Purity:
    ≥98% (HPLC)

Specification Test Results

About this item

 :
 :462.2 g/mol
Storage Temperature:Freezer
CAS Number:153660-03-8
Formula:C₁₀H₁₀BrN₅O₆PS·Na