Order Entry
United States
ContactUsLinkComponent
Granzyme B Assay Kit for Drug Discovery, Enzo Life Sciences
Granzyme B Assay Kit for Drug Discovery, Enzo Life Sciences
Catalog # 89156-542
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Granzyme B Assay Kit for Drug Discovery, Enzo Life Sciences
Catalog # 89156-542
Supplier:  Enzo Life Sciences
Supplier Number:  BML-AK711-0001
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    96 Wells
  • Storage Conditions:
    –80 °C (Long Term)
  • Application:
    Colorimetric Detection, Fluorescent Detection, HTS Activity Assay
  • Cat. No.:
    89156-542
  • Supplier no.:
    AK711-0001

Specifications

About this item

The Granzyme B Assay Kit for Drug Discovery is a complete assay system designed to measure the protease activity of granzyme B. It contains a colorimetric substrate (IEPD-pNA) to measure this activity; cleavage of the p-nitroanilide (pNA) group from the colorimetric substrate increases absorption at 405 nm.

The kit is useful to screen for inhibitors of granzyme B, a potential therapeutic target

Granzyme B is a serine protease found in the lymphocyte granules of cytotoxic T-lymphocytes (CTLs) and natural killer (NK) cells. Granzyme B plays a crucial role in the development of acute graft-vs.-host disease. Thus, the inhibition of this enzyme may prove to be an important tool in controlling immune responses.

An inhibitor 3, 4-Dichloroisocoumarin, is also included for use as a control.