ContactUsLinkComponent
Cathepsin K Drug Discovery Kit, Enzo Life Sciences
  89156-492
 :  
undefined
Cathepsin K Drug Discovery Kit, Enzo Life Sciences
  89156-492
 :  BML-AK430-0001
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Size:
    1 Kit
  • Cat. No.:
    89156-492
  • Supplier no.:
    AK430-0001
The Cathepsin K Drug Discovery Kit is a complete assay system designed to screen inhibitors of cathepsin K. Using a convenient microplate format, cathepsin K activity is monitored fluorimetrically using the substrate provided in the kit.
 : Quantity: 96 assays Contents: Cathepsin K (human) (recombinant), AMC substrate, AMC calibration standard, inhibitor, assay buffer, 1/2-volume white microplate.