Order Entry
United States
ContactUsLinkComponent
Matrix Metalloproteinase-2 (MMP-2) Colorimetric Drug Discovery Kit, Enzo Life Sciences
Matrix Metalloproteinase-2 (MMP-2) Colorimetric Drug Discovery Kit, Enzo Life Sciences
Catalog # 89156-466
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Matrix Metalloproteinase-2 (MMP-2) Colorimetric Drug Discovery Kit, Enzo Life Sciences
Catalog # 89156-466
Supplier:  Enzo Life Sciences
Supplier Number:  BML-AK408-0001
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    1 Kit
  • Storage Conditions:
    −80 °C
  • Application:
    Colorimetric Detection, HTS, Activity Assay
  • Cat. No.:
    89156-466
  • Supplier no.:
    AK408-0001

Specifications

About this item

A QuantiZymeTM Assay System

The MMP-2 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-2 (MMP-2, gelatinase A) using a thiopeptide as a chromogenic substrate. The assays are performed in a convenient 96-well microplate format. The kit is useful to screen inhibitors of MMP-2, a potential therapeutic target.

Delivery: Kit contains MMP-2 enzyme (human) (recombinant), substrate, inhibitor, assay buffer, 96-well microplate.