Order Entry
United States
ContactUsLinkComponent
CV-Caspase 3 & 7 Detection Kit, Enzo Life Sciences
CV-Caspase 3 & 7 Detection Kit, Enzo Life Sciences
Catalog # 89156-434
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
CV-Caspase 3 & 7 Detection Kit, Enzo Life Sciences
Catalog # 89156-434
Supplier:  Enzo Life Sciences
Supplier Number:  BML-AK118-0001
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    1 Kit
  • Storage Conditions:
    4 °C
  • Application:
    Fluorescence Microscopy, Fluorescent Detection
  • Cat. No.:
    89156-434
  • Supplier no.:
    AK118-0001

Specifications

About this item

Based on the dye cresyl violet and the caspase 3/7 DEVD peptide recognition sequence, this kit allows the detection of caspases 3 and 7 in living cells by fluorescence microscopy.

Once cells are grown, and apoptosis induced, staining is completed in just a few minutes

A consistant feature of mammalian cell apoptosis is the activation of caspases 3 and/or 7. When cleaved by caspase 3 or 7, the mono and unsubstituted cresyl violet fluoresce when excited at 550-590 nm.

Hoechst 33342 and Acridine Orange are also included to allow visualization of nuclei and lysosomes.

Delivery: Kit contains CR(DEVD)2 reagent, Acridine orange and Hoechst 33342 stain.