SCREEN-WELL® Kinase Inhibitor Library, Enzo Life Sciences
Supplier: Enzo Life Sciences
Certificates
About this item
The Screen-Well™ Kinase Inhibitor Library contains 80 known kinase inhibitors of well-defined activity. Inhibitors are supplied dissolved in DMSO at 10 mM and aliquoted into deep-well plates at either 100 or 500 microliters per well.
The library is an ideal tool for chemical genomics, assay development and other pharmacological applications. Includes inhibitors of these important kinases: Insulin/IGF Receptors, PI 3-Kinase, CaM Kinase II, JAK, PKA, CDK, JNK, PKC, CKI II, MAPK, RAF, EGFR, MEK, SAPK, GSK, MLCK, Src-family, IKK, PDGFR, VEGFR and many more.
Ordering information: Quantity: 100µl or 500µl per well
Contents: 80 compounds including inhibitors of these important kinases: BTK, CaM Kinase, CDK, CKI & II, EGFR, GSK, IKK, Insulin receptor, JAK, JNK, MAPK, MEK, MLCK, PI 3-Kinase, PDGFR, PKA, PKC, RAF, SAPK, Src-family, VEGFR, and more.
Contents: 80 compounds including inhibitors of these important kinases: BTK, CaM Kinase, CDK, CKI & II, EGFR, GSK, IKK, Insulin receptor, JAK, JNK, MAPK, MEK, MLCK, PI 3-Kinase, PDGFR, PKA, PKC, RAF, SAPK, Src-family, VEGFR, and more.
Specifications
- Concentration:DMSO solutions (10mM)