Order Entry
United States
ContactUsLinkComponent
Live-Dead Cell Staining Kit, Enzo Life Sciences
Live-Dead Cell Staining Kit, Enzo Life Sciences
Catalog # 89156-020
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Live-Dead Cell Staining Kit, Enzo Life Sciences
Catalog # 89156-020
Supplier:  Enzo Life Sciences
Supplier Number:  850-249-KI01
CAS Number:  

Specifications

  • Description:
    Live-Dead Cell Staining Kit
  • Size:
    1 kit
  • Cat. No.:
    89156-020
  • Supplier no.:
    850-249-KI01

Specifications

About this item

Kit utilizes Live-dye™, a cell permeable green fluorescent dye (Ex(max): 488nm; Em(max): 518nm), to stain live cells. Dead cells can be easily stained by propidium iodide (PI), a red fluorescent dye (Ex(max): 488nm; Em(max): 615nm), which in viable cells is actively pumped out of the cytoplasm.

  • Ready-to-use
  • Live-dead cell stain for 100 tests

Stained cells can be visualized by fluorescence microscopy using a band-pass filter (detects FITC and rhodamine). Cyto-dye has also been used to discriminate between apoptotic and non-apoptotic cells.