Order Entry
United States
ContactUsLinkComponent
Recombinant Caspase (active) set lll
Recombinant Caspase (active) set lll
Catalog # 89156-004
Supplier:  Enzo Life Sciences
CAS Number:  
Recombinant Caspase (active) set lll
Catalog # 89156-004
Supplier:  Enzo Life Sciences
Supplier Number:  850-243-KI01
CAS Number:  

Specifications

  • Enzyme type:
    Recombinant
  • Size:
    1 Set
  • Form:
    Lyophilized
  • Reconstitution Instructions:
    Reconstitute to 0.1 - 1 U/µl in PBS.
  • Enzyme Name:
    Caspase (active) set lll
  • Enzyme Activity:
    One unit of the recombinant human caspases is the enzyme activity that cleaves 1 nmol of the individual caspase substrate (YVAD-pNA for caspase-1, VDVAD-pNA for caspase-2, DEVD-pNA for caspase-3, VEID-pNA for caspase-6, DEVD-pNA for caspase-7, IETD-pNA fo
  • Purity:
    All components ≥90% (SDS-PAGE)
  • Cat. No.:
    89156-004
  • Supplier no.:
    850-243-KI01

Specifications

About this item

We recommend using 1 U per assay for analyzing caspase activity.

Active recombinant caspases are useful in screening caspase inhibitors, studying enzyme kinetics and regulation, determining substrate specificity, and as a positive control in caspase activity assays. 

Ordering information: Contents: Contains 25 units each of the following caspases:
- Caspase-1 (active) (human) (recombinant), yellow cap
- Caspase-2 (active) (human) (recombinant), orange cap
- Caspase-3 (active) (human) (recombinant), red cap
- Caspase-6 (active) (human) (recombinant), violet cap
- Caspase-7 (active) (human) (recombinant), red cap
- Caspase-8 (active) (human) (recombinant), green cap
- Caspase-9 (active) (human) (recombinant), blue cap
- Caspase-10 (active) (human) (recombinant), amber cap