ODN 2006 (Type B) (synthetic), TLRgrade™
Supplier: Enzo Life Sciences
Certificates
About this item
:
:7.720 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Source:Synthetic
- Formulation:Lyophilized. Sterile.
- Purity:Activity and endotoxin tested - TLRgrade™
- Endotoxin content:<0.002 EU/µg
- Quantity:128.9 nmol
Specification Test Results
Frequently Bought Together