Order Entry
United States
ContactUsLinkComponent
Toxin A
Toxin A
Catalog # 89152-986
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Toxin A
Catalog # 89152-986
Supplier:  Enzo Life Sciences
Supplier Number:  630-100-C100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Source:
  • Species:
  • Size:
    100 µg
  • Environmentally Preferable:
  • Protein/Peptide Name:
    Toxin A
  • Cat. No.:
    89152-986
  • Supplier no.:
    630-100-C100

Specifications

About this item

Cytotoxic and enterotoxic

Isolated from Clostridium difficile.

Has potent cytotoxic and enterotoxic properties. Translocates to the cytosol of target cells and glucosylates small GTP-binding proteins such as Rho, Rac, and Cdc42. Induces actin condensation, cell death and opening of the tight junctions. Triggers release of pro-inflammatory cytokines from intestinal epithelial cells probably via activation of MAP kinases.