Chlorotoxin
Catalog # 89152-962
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:500 µg
- Storage Conditions:Freezer
- Reconstitution Instructions:Reconstitute by injecting 1.4ml of distilled water into the vial using a calibrated syringe. Then dissolve the contents thoroughly to receive a 10-4 M solution.
- Protein/Peptide Name:Chlorotoxin
- Purity:>98% (TLC, HPLC)
- Cat. No.:89152-962
- Supplier no.:630-069-C500
Specifications
About this item
Potent neurotoxin. Small conductance Cl- channel blocker. Highly specific marker for glioma cells and tumors of neuroectodermal origin. Inhibits the activity of matrix metalloproteinase-2 (MMP-2).
- Cl- channel blocker
Lyophilized.
Soluble in water
Ordering information: Quantity: Vial contains exactly 0.55mg (0.14µmol)