

Lipopolysaccharides (from Salmonella minnesota), TLRgrade™
89152-782
:
- Size:2 ml
- Cat. No.:89152-782
- Supplier no.:581-018-L002
- Concentration:1 mg/ml
- Formulation:Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water
- Purity:Absence of detectable protein or DNA contaminants with agonistic TLR activity
:
:GHS07
:GHS08
:Refrigerator