Order Entry
United States
ContactUsLinkComponent
Lipopolysaccharides (from Salmonella minnesota), TLRgrade™
Health
Irritant
Lipopolysaccharides (from Salmonella minnesota), TLRgrade™
Catalog # 89152-780
Supplier:  Enzo Life Sciences
CAS Number:  
Health
Irritant
Lipopolysaccharides (from Salmonella minnesota), TLRgrade™
Catalog # 89152-780
Supplier:  Enzo Life Sciences
Supplier Number:  581-017-L002
CAS Number:  

Specifications

  • Size:
    2 ml
  • Cat. No.:
    89152-780
  • Supplier no.:
    581-017-L002

Specifications

Specification Test Results

  • Concentration:
    1 mg/ml
  • Formulation:
    Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water
  • Purity:
    Absence of detectable protein or DNA contaminants with agonistic TLR activity

Specification Test Results

About this item

 :
 :GHS08
 :GHS07
Storage Temperature:Refrigerator