5-(N-Ethyl-N-isopropyl)amiloride ≥98% (by TLC)
Catalog # 89152-424
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:5 mg
- Cat. No.:89152-424
- Supplier no.:550-266-M005
Specifications
About this item
:
:299.763 g/mol
Storage Temperature:Refrigerator
CAS Number:1154-25-2
Formula:C₁₁H₁₈ClN₇O
MDL Number:MFCD00151710