Human Recombinant beta-Nerve Growth Factor (from E. coli)
Catalog # 89151-660
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:5 µg
- Biological Activity:Neurotrophic activity was bioassayed using rat pheochromocytoma PC12 cell cultures over a period of 5-14 days. Activity is in the range of 1-50ng/ml.
- Reconstitution Instructions:Reconstitue in water or DMEM to a final concentration of at least 10µg/ml and store at -80°C before use.
- Protein/Peptide Name:beta-Nerve Growth Factor
- Purity:>98% (HPLC)
- Formulation:Lyophilized. No additives.
- Cat. No.:89151-660
- Supplier no.:521-005-C005
Specifications
About this item
Produced in E. coli. Dimer of two identical subunits (119 aa) linked together by very strong hydrophobic interactions.
Ordering information: Quantity: 5 µg (determined by amino acid analysis).