Order Entry
United States
ContactUsLinkComponent
 
Ergocalciferol (vitamin D2, calciferol) ≥98% (by HPLC)
Catalog # 89151-440
Supplier:  Avantor
CAS Number:  
undefined
Ergocalciferol (vitamin D2, calciferol) ≥98% (by HPLC)
Catalog # 89151-440
Supplier:  Avantor
CAS Number:  

Specifications

  • Size:
    1 g
  • Cat. No.:
    89151-440
  • Supplier no.:
    460-025-G001

Specifications

About this item

 :
ADR:6.1,II
 :396.66 g/mol
UN:2811
CAS Number:50-14-6
Formula:C₂₈H₄₄O
MDL Number:MFCD00166988