R-Sulforaphane ≥98% (by UHPLC)
Catalog # 89149-294
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:10 mg
- Cat. No.:89149-294
- Supplier no.:350-230-M010
Specifications
About this item
:
:177.2876 g/mol
CAS Number:142825-10-3
Formula:C₆H₁₁NOS₂
MDL Number:MFCD09039283