Order Entry
United States
ContactUsLinkComponent
PQ401 ≥98% (by HPLC and UV method)
PQ401 ≥98% (by HPLC and UV method)
  89147-962
 :  
undefined
PQ401 ≥98% (by HPLC and UV method)
  89147-962
 :  270-459-M005
 :  

 

  • Size:
    5 mg
  • Cat. No.:
    89147-962
  • Supplier no.:
    270-459-M005

 

 

 :
 :341.8 g/mol
 :Freezer
 :196868-63-0
 :C₁₈H₁₆ClN₃O₂
 :MFCD00160558