- Antibody Type:Primary
- Antigen Name:Peroxisomal Membrane Protein 70
- Antigen Symbol:Peroxisomal Membrane Protein 70
- Clonality:Polyclonal
- Flow Cytometry:Yes
- Host:Rabbit
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid. In PBS containing 1mg/ml BSA and 0.05% sodium azide.
- Concentration:1mg/ml
- Purification:Epitope-affinity purified IgG
- Cat. No.:89145-306
- Supplier no.:210-205-C100
Peroxisomes are single membrane organelles which are involved in many important biochemical pathways and are found in almost all eukaryotic cells. Various toxins including phenols, alcohols, and aldehydes are targeted for modification by peroxisomes. Long chain fatty acid metabolism via β-oxidation is also mediated by this organelle. Loss of peroxisomal functions may result in diseases such as Zellweger syndrome, hyperpipecolic acidemia, neonatal adrenoleukodystrophy, and infantile Refsums disease. Peroxisomal membrane protein 70 (PMP70) is an abundant, integral membrane protein of the peroxisome. This protein is induced by treatment with hypolipidemic agents in parallel with peroxisome proliferation and stimulation of the peroxisomal β-oxidation enzymes.
- For Western Blot
Recognizes mouse PMP70. Detects a band of ~70kDa by Western blot.
Type: Primary
Antigen: PEROXISOMAL MEMBRANE PROTEIN 70
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse