Human Recombinant PTX3 (from CHO Cells)
Catalog # 89144-128
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Human
- Size:10 µg
- Protein/Peptide Name:PTX3
- Concentration:0.1mg/ml.
- Formulation:Liquid. In PBS containing 2% BSA and 0.1% sodium azide.
- Cat. No.:89144-128
- Supplier no.:201-124-C010
Specifications
About this item
Produced in CHO cells. Human full-length PTX3.