Order Entry
United States
ContactUsLinkComponent
1-Methyl-L-tryptophan ≥95%
1-Methyl-L-tryptophan ≥95%
Catalog # 89142-916
Supplier:  Avantor
CAS Number:  
undefined
1-Methyl-L-tryptophan ≥95%
Catalog # 89142-916
Supplier:  Avantor
CAS Number:  

Specifications

  • Size:
    50 mg
  • Cat. No.:
    89142-916
  • Supplier no.:
    106-040-M050

Specifications

About this item

 :
 :218.3 g/mol
Storage Temperature:Ambient
CAS Number:21339-55-9
Formula:C₁₂H₁₄N₂O₂