To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Host:
- Primary antibody reactivity:Human
- Description:Heat shock transcription factor 1 ELISA kit
- Size:96 wells
- Environmentally Preferable:
- Storage Conditions:+4 °C (long term), -20 °C (for standards and microtiter plate)
- Detection Method:Colorimetric
- Sensitivity:35 pg/mL (Range 0.39 - 12.5 ng/mL)
- Cat. No.:89135-344
- Supplier no.:900-198
Specifications
About this item
The HSF1 EIA kit is a complete kit for the quantitative determination of HSF1 in cell lysates and tissue extracts from human and mouse origins
- Use less precious sample compared to Western blot
- Ready to use liquid color-coded reagents and pre-coated plate reduces error
For the quantitative determination of human and mouse HSF1 in cell lysates and tissue.
This is a Sensitive ELISA kit for this key transcription factor for heat shock protein (chaperone) research.