Cell lysis buffer 4, (100 ml), Enzo Life Sciences
Catalog # 95043-744
Supplier: Enzo Life Sciences
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Size:100 mL
- Formulation:Liquid. 100mM PBS, pH 7.3, containing 150mM sodium chloride, 1mM EDTA, 0.5% Triton X-100 and 0.005% Tween 20.
- Cat. No.:95043-744
- Supplier no.:80-1339