To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Description:Benchtop Water Quality Meter
- Environmentally Preferable:
- Dimensions:217 × 168 × 58 mm (8.5 × 6.5 × 2.25")
- Weight:533 g (18 oz.)
- Cat. No.:97039-652
- Supplier no.:860033
Reads pH, ORP (oxygen reduction potential), mV, conductivity, TDS (total dissolved solids), salinity and temperature.
Rugged housing with large LCD displaying the parameter being measured together with time, date and temperature (in °C or °F). Measures: ph, ORP (oxygen reduction potential - SOLD SEPARATELY), mV, Conductivity, TDS (total dissolved solids), Salinity and Temperature. Features: automatic or manual temperature compensation, automatic or manual ranging, automatic buffer recognition, digital and analog outputs, min/max and 99 data points, data hold, 5 calibration points, and electrode status indication.
Operates on AC voltage (no batteries). Also accepts any pH or ORP probe with a standard BNC connector.